missing translation for 'onlineSavingsMsg'
Learn More

Iodoform, 99%

Catalog No. AAA1366430
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1366430 250 g
AAA1366418 50 g
AAA136640B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1366430 Supplier Thermo Scientific Chemicals Supplier No. A1366430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75-47-8 | CHI3 | 393.732 g/mol

Iodoform is use as a disinfectant. It is an active ingredient of ear powder along with other constituents like zinc oxide and propanoic acid, which prevent infection and facilitate the removal of ear hair in dogs and cats. In analytical chemistry, it is involved in the determination of iodoform in drinking water as well as to remove heavy metals like mercury from fluids. It plays an important role in the fermentation of bio mass. It is also used in root canal therapy for infected primary teeth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoform is use as a disinfectant. It is an active ingredient of ear powder along with other constituents like zinc oxide and propanoic acid, which prevent infection and facilitate the removal of ear hair in dogs and cats. In analytical chemistry, it is involved in the determination of iodoform in drinking water as well as to remove heavy metals like mercury from fluids. It plays an important role in the fermentation of bio mass. It is also used in root canal therapy for infected primary teeth.

Solubility
Soluble in water, ether, acetic acid, ethanol, chloroform, ether, glycerol, carbon disulfide, olive oil, benzene and acetone. Slightly soluble in petroleum ether.

Notes
Light sensitive. Incompatible with strong bases, alkali metals, trong oxidizing agents and magnesium.

Chemical Identifiers

CAS 75-47-8
Molecular Formula CHI3
Molecular Weight (g/mol) 393.732
MDL Number MFCD00001069
InChI Key OKJPEAGHQZHRQV-UHFFFAOYSA-N
Synonym triiodomethane, methane, triiodo, carbon triiodide, jodoform, trijodmethane, dezinfekt v, jodoform czech, trijodmethane czech, ccris 346, unii-kxi2j76489
PubChem CID 6374
ChEBI CHEBI:37758
IUPAC Name iodoform
SMILES C(I)(I)I

Specifications

Melting Point 118°C to 123°C
Density 4.008
Flash Point None
Odor Unpleasant
Quantity 250 g
Beilstein 1697010
Sensitivity Light sensitive
Merck Index 14,5033
Solubility Information Soluble in water,ether,acetic acid,ethanol,chloroform,ether,glycerol,carbon disulfide,olive oil,benzene and acetone. Slightly soluble in petroleum ether.
Formula Weight 393.73
Percent Purity 99%
Chemical Name or Material Iodoform
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 200-874-5
RTECSNumber PB7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only