missing translation for 'onlineSavingsMsg'
Learn More

Iodoform, 99+%

Catalog No. AC149125000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149125000 500 g Glass bottle
AC149121000 100 g Glass bottle
AC149120025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC149125000 Supplier Thermo Scientific Chemicals Supplier No. 14912 5000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75-47-8 | CHI3 | 393.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-47-8
Molecular Formula CHI3
Molecular Weight (g/mol) 393.72
MDL Number MFCD00001069
InChI Key OKJPEAGHQZHRQV-UHFFFAOYSA-N
Synonym triiodomethane, methane, triiodo, carbon triiodide, jodoform, trijodmethane, dezinfekt v, jodoform czech, trijodmethane czech, ccris 346, unii-kxi2j76489
PubChem CID 6374
ChEBI CHEBI:37758
IUPAC Name iodoform
SMILES C(I)(I)I

Specifications

Melting Point 119.0°C to 122.0°C
Color Yellow
Ignition Residue 0.1% max.
Loss on Drying 0.5% max. (24 hrs) (over silicagel)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CHI3
Quantity 500 g
Beilstein 01, 73
Fieser 03,165; 04,266
Merck Index 15, 5076
Solubility Information Solubility in water: 0.1g/L (20°C). Other solubilities: 1g/60mL cold alcohol- 1g/16mL boiling alcohol,1g/10mL chloroform- 1g/7.5mL ether,1g/80mL glycerol- 1g/3mL cs2,freely soluble in benzene and acetone
Formula Weight 393.72
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Iodoform, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-874-5
RTECSNumber PB7000000
TSCA TSCA

RUO – Research Use Only