missing translation for 'onlineSavingsMsg'
Learn More

Iodoethane (stabilized with Copper chip) 99.0+%, TCI America™

Supplier:  TCI America I0058500G

 View more versions of this product

Catalog No. I0058500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Iodoethane (stabilized with Copper chip)
-108°C
72°C
MFCD00001091
1993
HVTICUPFWKNHNG-UHFFFAOYSA-N
iodoethane
6340
155.97
Liquid
75-03-6
Colorless
C2H5I
500 g
ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czech
CCI
155.966
CHEBI:42487
≥99.0% (GC)

Chemical Identifiers

75-03-6
155.966
HVTICUPFWKNHNG-UHFFFAOYSA-N
6340
iodoethane
C2H5I
MFCD00001091
ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czech
CHEBI:42487
CCI
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0079

RTECSNumber : KI4750000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions