Learn More
Iodoethane, 98+%, stab. with copper
CAS: 75-03-6 | C2H5I | 155.966 g/mol
Supplier: Thermo Scientific Chemicals A144440E
Description
Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.
Solubility
Miscible with water.
Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and magnesium.
Specifications
| Iodoethane | |
| 75-03-6 | |
| 1.95 | |
| >73°C (163°F) | |
| 1.512 | |
| 2500 g | |
| Light and moisture sensitive | |
| ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czech | |
| HVTICUPFWKNHNG-UHFFFAOYSA-N | |
| iodoethane | |
| 6340 | |
| 155.97 |
| stabilized with copper | |
| -108°C | |
| 70°C to 73°C | |
| C2H5I | |
| MFCD00001091 | |
| 505934 | |
| 14,3813 | |
| Miscible with water. | |
| CCI | |
| 155.966 | |
| CHEBI:42487 | |
| ≥98% |
Chemical Identifiers
| 75-03-6 | |
| 155.966 | |
| HVTICUPFWKNHNG-UHFFFAOYSA-N | |
| 6340 | |
| iodoethane |
| C2H5I | |
| MFCD00001091 | |
| ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czech | |
| CHEBI:42487 | |
| CCI |
Safety and Handling
GHS H Statement
H334-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P210-P235-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
H227-H302-H315-H317-H319-H334-H335
EINECSNumber : 200-833-1
RTECSNumber : KI4750000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only