missing translation for 'onlineSavingsMsg'
Learn More

Iodoethane, 98+%, stab. with copper

Catalog No. AAA1444436
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1444436 500 g
AAA1444422 100 g
AAA144440E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1444436 Supplier Thermo Scientific Chemicals Supplier No. A1444436

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75-03-6 | C2H5I | 155.966 g/mol

Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and magnesium.

Chemical Identifiers

CAS 75-03-6
Molecular Formula C2H5I
Molecular Weight (g/mol) 155.966
MDL Number MFCD00001091
InChI Key HVTICUPFWKNHNG-UHFFFAOYSA-N
Synonym ethyl iodide, ethane, iodo, monoiodoethane, hydriodic ether, ethyliodide, jodethan czech, ethyljodid, jodethan, iodo-ethane, ethyljodid czech
PubChem CID 6340
ChEBI CHEBI:42487
IUPAC Name iodoethane
SMILES CCI

Specifications

Melting Point -108°C
Density 1.95
Boiling Point 70°C to 73°C
Flash Point >73°C (163°F)
Refractive Index 1.512
Quantity 500 g
Beilstein 505934
Sensitivity Light and moisture sensitive
Merck Index 14,3813
Solubility Information Miscible with water.
Formula Weight 155.97
Percent Purity ≥98%
Chemical Name or Material Iodoethane, stabilized with copper
Show More Show Less
Hazard Category H227-H302-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
EINECSNumber 200-833-1
RTECSNumber KI4750000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only