missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetonitrile 98.0+%, TCI America™

Catalog No. p-7131399
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04195G 5 g
I041925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04195G Supplier TCI America Supplier No. I04195G
Only null left

Chemical Identifiers

CAS 624-75-9
Molecular Formula C2H2IN
Molecular Weight (g/mol) 166.949
MDL Number MFCD00001886
InChI Key VODKOOOHHCAWFR-UHFFFAOYSA-N
Synonym iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
PubChem CID 69356
IUPAC Name 2-iodoacetonitrile
SMILES C(C#N)I

Specifications

Color Brown-Yellow
Boiling Point 78°C
Quantity 5 g
UN Number 1760
Formula Weight 166.95
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Iodoacetonitrile