missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetonitrile, 97%

Catalog No. AAL0363418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0363418 50 g
AAL0363409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0363418 Supplier Thermo Scientific Chemicals Supplier No. L0363418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 624-75-9 | C2H2IN | 166.949 g/mol

Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetonitrile is used for alkylation of nucleophiles. It undergoes Reformatsky and Wittig-type reactions with aldehydes.

Solubility
Soluble in benzene, alcohol, acetone, ether. Slightly soluble in water. Insoluble in hexane.

Notes
Light Sensitive. Store at 4°C. Incompatible with reducing agents, light and bases. Store away from strong bases.

Chemical Identifiers

CAS 624-75-9
Molecular Formula C2H2IN
Molecular Weight (g/mol) 166.949
MDL Number MFCD00001886
InChI Key VODKOOOHHCAWFR-UHFFFAOYSA-N
Synonym iodoacetonitrile, acetonitrile, iodo, iodo-acetonitrile, 2-iodoethanenitrile, 2-iodo acetonitrile, 2-iodoaceto-nitrile, 2-iodanylethanenitrile, pubchem12543, acmc-1ba3n
PubChem CID 69356
IUPAC Name 2-iodoacetonitrile
SMILES C(C#N)I

Specifications

Density 2.306
Boiling Point 70°C to 72°C (10 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.574
Linear Formula ICH2CN
Quantity 50 g
UN Number UN2922
Beilstein 1734628
Sensitivity Light sensitive
Solubility Information Soluble in benzene,alcohol,acetone,ether. Slightly soluble in water. Insoluble in hexane.
Formula Weight 166.95
Percent Purity 97%
Chemical Name or Material Iodoacetonitrile
Show More Show Less
Hazard Category H227-H301+H311+H331-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 210-860-0
RTECSNumber AM0740000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only