missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetamide Crystalline MP Biomedicals

Catalog No. ICN10035105
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10035105 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10035105 Supplier MP Biomedicals Inc Supplier No. 0210035105

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144-48-9
Molecular Formula C2H4INO
Molecular Weight (g/mol) 184.964
MDL Number MFCD00008028
InChI Key PGLTVOMIXTUURA-UHFFFAOYSA-N
Synonym iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid
PubChem CID 3727
IUPAC Name 2-iodoacetamide
SMILES C(C(=O)N)I

Specifications

Color Tan
Quantity 5 g
Formula Weight 185
Physical Form Powder
Chemical Name or Material Iodoacetamide, Molecular biology and Cell biology
EINECSNumber 205-630-1
Recommended Storage Store at Room Temperature -0°C