missing translation for 'onlineSavingsMsg'
Learn More

Iodine, specified according to the requirements of Ph.Eur, BP, USP

CAS: 7553-56-2 | I2 | 253.81 g/mol

$192.30 - $192.30

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC457762500
View Documents
Thermo Scientific Chemicals
457762500
250 g
Each for $192.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7553-56-2
253.81
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
diiodine
I2
MFCD00011355 MFCD00164163
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
CHEBI:17606
II

Specifications

185.0°C
7553-56-2
I2
MFCD00011355 MFCD00164163
Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in aq.sol. of hi or iodines, soluble in alcohol, carbon disulfide, chloroform,, ether, ccl4 and glycerol
II
253.81
CHEBI:17606
99.8 to 100.5% (Iodometry)
113.0°C
250 g
I2
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PNDPGZBMCMUPRI-UHFFFAOYSA-N
diiodine
807
253.81
Iodine, specified according to the requirements of Ph.Eur, BP, USP
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Causes damage to organs through pr

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 231-442-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only