missing translation for 'onlineSavingsMsg'
Learn More

Iodine, specified according to the requirements of Ph.Eur, BP, USP

Catalog No. AC457762500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457762500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC457762500 Supplier Thermo Scientific Chemicals Supplier No. 457762500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7553-56-2 | I2 | 253.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Boiling Point 185.0°C
Melting Point 113.0°C
Quantity 250 g
Linear Formula I2
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in aq.sol. of hi or iodines, soluble in alcohol, carbon disulfide, chloroform,, ether, ccl4 and glycerol
Formula Weight 253.81
Percent Purity 99.8 to 100.5% (Iodometry)
Chemical Name or Material Iodine, specified according to the requirements of Ph.Eur, BP, USP
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Causes damage to organs through pr
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and kee
EINECSNumber 231-442-4

RUO – Research Use Only