missing translation for 'onlineSavingsMsg'
Learn More

Iodine, resublimed crystals, ACS, 99.8%

Catalog No. AA4195536
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4195536 500 g
AA4195522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AA4195536 Supplier Thermo Scientific Chemicals Supplier No. 04195536
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 7553-56-2 | I2 | 253.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II

Specifications

Melting Point 183°C to 186° (sublimation)
Color Gray-Purple
Physical Form Prills or Crystals
Quantity 500 g
Assay Percent Range 99.8%
Formula Weight 253.81
Grade ACS
Odor Irritating
Appearance Bluish-black
Density 4.93
Chemical Name or Material Iodine
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335-H372
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-442-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only