missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, puriss., Honeywell Fluka™
Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.8-100.5%
$996.88 - $996.88
Chemical Identifiers
| CAS | 7553-56-2 |
|---|---|
| Molecular Formula | I2 |
| Molecular Weight (g/mol) | 253.81 |
| MDL Number | MFCD00011355 MFCD00164163 |
| InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
| Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
| PubChem CID | 807 |
| ChEBI | CHEBI:17606 |
| SMILES | II |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7553-56-2 | |
| 253.81 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| 807 | |
| II |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| CHEBI:17606 |
Specifications
| 7553-56-2 | |
| I2 | |
| 3495 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| II | |
| 253.81 | |
| CHEBI:17606 | |
| 99.8 to 100.5% | |
| 0.31mmHg (25°C), 1mmHg (38.7°C) | |
| Iodine, Meets analytical specification of Ph. Eur.BP, USP |
| 500 g | |
| MFCD00011355 MFCD00164163 | |
| 3587194 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| molecular iodine | |
| 807 | |
| 253.81g/mol | |
| puriss. | |
| Vapor Density: 9 (vs air) |
Safety and Handling
P261-P273-P280-P305 + P351 + P338-P314
H312 + H332-H315-H319-H335-H372-H400
EINECSNumber : 231-442-4
RTECSNumber : NN1575000