missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, Puriss., ≥99.5% (RT), particles (round), Honeywell Fluka™
Puriss.,≥99.5% (RT), particles (round)
$1879.85 - $1879.85
Chemical Identifiers
| CAS | 7553-56-2 |
|---|---|
| Molecular Formula | I2 |
| Molecular Weight (g/mol) | 253.81 |
| MDL Number | MFCD00011355 MFCD00164163 |
| InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
| Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
| PubChem CID | 807 |
| ChEBI | CHEBI:17606 |
| SMILES | II |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7553-56-2 | |
| 253.81 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| 807 | |
| II |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| CHEBI:17606 |
Specifications
| 184°C (literature) | |
| 7553-56-2 | |
| 1 kg | |
| I2 | |
| UN3495 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| II | |
| 253.81 | |
| CHEBI:17606 | |
| ≥99.5% | |
| Glass Bottle | |
| Iodine, Puriss., ≥99.5% (RT), particles (round) |
| 113°C (literature) | |
| 100.0000% | |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| 3587194 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| diiodine | |
| 807 | |
| 253.81 | |
| puriss. | |
| 4.930 g/cm3 |
Safety and Handling
ShelfLife : 1620 days from date of manufacture
DOTInformation : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Iodine
EINECSNumber : 231-442-4
Recommended Storage : Room Temp