missing translation for 'onlineSavingsMsg'
Learn More

Iodine monochloride

Catalog No. p-4407451
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3910414 25 g
AA3910422 100 g
AA3910436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3910414 Supplier Thermo Scientific Chemicals Supplier No. 03910414
Only null left

CAS: 7790-99-0 | ClI | 162.35 g/mol

ICl is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Boiling Point 96°C to 98°C
Melting Point 25°C to 27°C
Physical Form Crystalline, Liquid
Quantity 25 g
Beilstein 3902972
Solubility Information Soluble in water,alcohol,ether,carbon disulfide.
Formula Weight 162.36
Sensitivity Light and moisture sensitive
Density 3.1822 g/mL
Chemical Name or Material Iodine monochloride
Hazard Category H300-H311-H314-H335-H373
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-236-7
RTECSNumber NN1650000
TSCA Yes
storageNote1 Moisture Sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only