missing translation for 'onlineSavingsMsg'
Learn More

Iodine Monochloride Solution WIJS AOAC, Reagents

Catalog No. 361059155
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361059155 4 L
361059147 1 L
361059152 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
DEA LIST I
Catalog No. 361059155 Supplier Labchem Inc Supplier No. C72181004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DEA LIST I
  • Iodine Monochloride Solution, WIJS
  • For Determining Iodine Absorption Number of Fat and Oils
  • Appearance: Brown liquid
  • Specific Gravity: 1.05
  • Boiling Point: 118°C
  • Brand: Reagents
  • Iodine/Chlorine Ratio Spec: 1.05 to 1.15
  • Total Halogen Content (vs Sodium Thiosulfate/Starch) Spec: 0.189 to 0.193 N at 20°C

Specifications

Chemical Name or Material Iodine Monochloride Solution, WIJS
Color Blend
CAS 7553-56-2, 64-19-7, 7790-99-0
Physical Form Liquid or Solid
Quantity 4 L
Molecular Weight (g/mol) Blend
Percent Purity 100
Grade Laboratory
Concentration or Composition (by Analyte or Components) Amber Glass Bottle
Packaging Amber Glass Bottle