missing translation for 'onlineSavingsMsg'
Learn More

Iodine monochloride, approx. 0.22N soln. in glacial acetic acid

Catalog No. p-4407449
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42647AP 500 mL
AA42647K2 1 L
AA42647K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42647AP Supplier Thermo Scientific Chemicals Supplier No. 042647AP
Only null left

For Determination of Iodine Absorption Number of Fats | CAS: 7790-99-0 | ClI | 162.35 g/mol

For determination of iodine absorption number of fatsIodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture and Light Sensitive

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Physical Form Liquid
Quantity 500 mL
Solubility Information Miscible with water,alcohol,ether and carbon disulfide.
Formula Weight 162.36
Ignition Point 485°C (905°F) (acetic acid)
Sensitivity Light and moisture sensitive
Appearance Liquid
Chemical Name or Material Iodine monochloride
Hazard Category H226-H314-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8, Packing Group: II
EINECSNumber 232-236-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only