missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine monochloride, approx. 0.22N soln. in glacial acetic acid
Description
For determination of iodine absorption number of fatsIodine monochloride is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Moisture and Light Sensitive
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 4 L |
| Ignition Point | 485°C (905°F) (acetic acid) |
| Sensitivity | Light and moisture sensitive |
| Appearance | Liquid |
| Solubility Information | Miscible with water,alcohol,ether and carbon disulfide. |
| Formula Weight | 162.36 |
| Chemical Name or Material | Iodine monochloride |
Safety and Handling
| Hazard Category | H226-H314-H335 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 8, Packing Group: II |
| EINECSNumber | 232-236-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only