missing translation for 'onlineSavingsMsg'
Learn More

Iodine monochloride, ACS reagent

Catalog No. p-43815
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412361000 100 g Glass bottle
AC412365000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC412361000 Supplier Thermo Scientific Chemicals Supplier No. 41236 1000
Only null left

CAS: 7790-99-0 | ClI | 162.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Boiling Point 97.0°C
Melting Point 26.0°C
Color Brown-Red
Physical Form Fuming Liquid
Quantity 100 g
Linear Formula ICl
Fieser 01,502; 08,261; 10,212; 11,268
Merck Index 15, 5060
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes, soluble in ethanol, ether and cs2
Formula Weight 162.36
Percent Purity 1.0 to 1.2 (I/Cl ratio of the WIJS solution prepared from the reagent)
Grade ACS Reagent
Packaging Glass bottle
Specific Gravity 3.2
Density 3.2000g/mL
Chemical Name or Material Iodine monochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Fatal if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 232-236-7
RTECSNumber NN1650000
TSCA TSCA

RUO – Research Use Only