missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Iodine lump, ultra dry, 99.999% (metals basis)
Specifications
Specifications
| Melting Point | 183-186° subl. |
| CAS | 7553-56-2 |
| Physical Form | Solid |
| Quantity | 5 g |
| Density | 4.930 |
| Molecular Formula | I{2} |
| MDL Number | MFCD00011355 |
| UN Number | UN3495 |
| Formula Weight | 253.81 |
| Percent Purity | 99.999% |