missing translation for 'onlineSavingsMsg'
Learn More

Iodine (Iodine-Iodide), 0.315 N, Ricca Chemical

Catalog No. R40120004C
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R40120004C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R40120004C Supplier Ricca Chemical Company Supplier No. R40120004C
Only null left

Specifications

Chemical Name or Material Iodine
Boiling Point 100°C
Melting Point 0°C
CAS 7553-56-2
CAS Min % 3.48, 10.95, 85.57
CAS Max % 3.48, 10.95, 85.57
Quantity 4 L
Assay Percent Range 0.3148 to 0.3152
Assay Percent Notes Normality Iodine
Packaging Amber Glass Bottle
Appearance Dark Brown Liquid
Show More Show Less