missing translation for 'onlineSavingsMsg'
Learn More

Iodine, D'Antoni, Ricca Chemical

$238.56 - $335.06

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
410316
View Documents
Ricca Chemical Company
410316
500 mL Amber Glass Bottle
Each for $238.56
Only null left
 
410332
View Documents
Ricca Chemical Company
410332
1 L Amber Glass Bottle
Each for $335.06
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7553-56-2
Mixture
PNDPGZBMCMUPRI-UHFFFAOYSA-N
807
diiodine
I2
MFCD00011355 MFCD00164163
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
CHEBI:17606
II

Specifications

7553-56-2,7732-18-5,7681-11-0
1.0,100.0,1.0
MFCD00011355 MFCD00164163
PNDPGZBMCMUPRI-UHFFFAOYSA-N
diiodine
807
Laboratory
Dark Brown
Liquid
0.96,96.0,0.96
I2
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
II
Mixture
CHEBI:17606
Amber Glass Bottle
500 mL
Iodine, D'Antoni
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'