Learn More
Iodine, crystalline, 99.99+% (metals basis)
CAS: 7553-56-2 | I2 | 253.81 g/mol
Supplier: Thermo Scientific Chemicals 00015830
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Iodine, Crystalline | |
| 7553-56-2 | |
| 250 g | |
| I2 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| II | |
| 253.81 | |
| CHEBI:17606 | |
| 99.9% |
| 183°C to 186°C (sublimation) | |
| Solid | |
| >99.99% (Metal Basis) | |
| MFCD00011355 MFCD00164163 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| molecular iodine | |
| 807 | |
| 253.81 | |
| 4.93 g/cm3 |
Chemical Identifiers
| 7553-56-2 | |
| 253.81 | |
| PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
| 807 | |
| II |
| I2 | |
| MFCD00011355 MFCD00164163 | |
| iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
| CHEBI:17606 |
Safety and Handling
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H372
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 231-442-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only