missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine-Bromine Solution, Hanus (AOAC) for Iodine Absorption Number of Fats and Oils, Reagents
Iodine-Bromine Solution, Hanus (AOAC) for Iodine Absorption Number of Fats and Oils, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$327.83 - $568.70
Specifications
| Boiling Point | 118°C |
|---|---|
| Chemical Name or Material | Iodine-Bromine Solution, Hanus (AOAC) for Iodine Absorption Number of Fats and Oils |
| CAS | 64-19-7,7553-56-2,7553-56-2 |
| Color | Brown |
| pH | <2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-336-476
|
Labchem Inc
C72180001C |
1 L |
Each for $568.70
|
|
|||||
|
01-336-475
|
Labchem Inc
C7218000500C |
500 mL |
Each for $327.83
|
|
|||||
Specifications
| 118°C | |
| 64-19-7,7553-56-2,7553-56-2 | |
| <2 | |
| Reagent | |
| 1.07 |
| Iodine-Bromine Solution, Hanus (AOAC) for Iodine Absorption Number of Fats and Oils | |
| Brown | |
| Liquid | |
| Glass Bottle |