missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 99.8%, ACS DEA List I, Reagents

Catalog No. 361059157
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
12 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361059157 100 g
361059156 12 kg
361059158 25 g
361059161 2 kg
361059163 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
DEA LIST I
Catalog No. 361059157 Supplier Labchem Inc Supplier No. C1217500100C2
DEA LIST I
  • Iodine, ACS Grade
  • Brand: Reagents
  • DEA List I
  • Appearance: Violet-black Crystals
  • Molecular Formula I2
  • MW (g/mol): 253.8089
  • Assay >= 99.8 %
  • Chlorine and Bromine (as Cl) <= 0.005 %
  • Nonvolatile matter <= 0.01 %

Chemical Identifiers

Molecular Weight (g/mol) 253.81

Specifications

Physical Form Crystals and/or Chunks
Quantity 100 g
Percent Purity 99.80%
Grade ACS