missing translation for 'onlineSavingsMsg'
Learn More

Iodine 0.315 Normal DEA List I, Reagents

Catalog No. p-200104729
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361059171 1 L
361059167 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
DEA LIST I
Catalog No. 361059171 Supplier Labchem Inc Supplier No. CI1004001C
Only null left
DEA LIST I
  • Iodine 0.315 N
  • DEA List I
  • Appearance: Dark Brown Liquid
  • Specific Gravity: 1.15
  • Brand: Reagents
  • NIST Traceable
  • Prepared with ACS Grade Components and ASTM Type II Water
  • Assay (vs. Sodium Thiosulfate/Starch) Spec: 0.3145 to 0.3155 N

Chemical Identifiers

Molecular Weight (g/mol) Blend

Specifications

Physical Form Liquid or Solid
Quantity 1 L
Percent Purity 0.315 N
Grade Laboratory