missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine 0.3125 Normal DEA List I, Reagents
Description
- Iodine 0.3125 N
- DEA List I
- Appearance: Dark Brown Liquid
- Specific Gravity: 1.15
- Brand: Reagents
- NIST Traceable
- Prepared with ACS Grade Components and ASTM Type II Water
Specifications
Specifications
| Physical Form | Liquid or Solid |
| Quantity | 1 L |
| Percent Purity | 0.3125 N |
| Grade | Laboratory |