missing translation for 'onlineSavingsMsg'
Learn More

Iodine 0.3125 Normal DEA List I, Reagents

Catalog No. 361059165
Change view
Click to view available options
Quantity:
1 L
2.5 L
4L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361059165 1 L
361059159 2.5 L
361059170 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
DEA LIST I
Catalog No. 361059165 Supplier Labchem Inc Supplier No. CI1006001C
DEA LIST I
  • Iodine 0.3125 N
  • DEA List I
  • Appearance: Dark Brown Liquid
  • Specific Gravity: 1.15
  • Brand: Reagents
  • NIST Traceable
  • Prepared with ACS Grade Components and ASTM Type II Water

Chemical Identifiers

Molecular Weight (g/mol) Blend

Specifications

Physical Form Liquid or Solid
Quantity 1 L
Percent Purity 0.3125 N
Grade Laboratory