missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 0.08 Normal in Methanol, Reagents

Catalog No. p-200021721
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334914 1 L
01334913 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334914 Supplier Labchem Inc Supplier No. CI2018001C
Only null left

Iodine, 0.08 Normal in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 67-56-1 , 7553-56-2
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Diiodine solution
IUPAC Name methanol
SMILES CO

Specifications

Boiling Point 64.5°C
Color Dark Brown
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8
Chemical Name or Material Iodine, 0.08 Normal in Methanol