missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 0.08 Normal in Methanol, Reagents

Catalog No. 01334914
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334914 1 L
01334913 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334914 Supplier Labchem Inc Supplier No. CI2018001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Iodine, 0.08 Normal in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 67-56-1 , 7553-56-2
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Diiodine solution
IUPAC Name methanol
SMILES CO

Specifications

Boiling Point 64.5°C
Color Dark Brown
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8
Chemical Name or Material Iodine, 0.08 Normal in Methanol