missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 0.002 Normal, Unstandardized, Reagents

Catalog No. 01334909
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334909 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334909 Supplier Labchem Inc Supplier No. CI2120004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Iodine, 0.002 Normal, Unstandardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Iodine, 0.002 Normal, Unstandardized
Boiling Point 100°C
CAS 7732-18-5, 7681-11-0
Color Dark Brown
Physical Form Liquid
Quantity 4 L
Molecular Formula KI/I2/H2O
Synonym Diiodine solution
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.001
Show More Show Less