missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodide Iodate concentrate, For 1L standard solution, 0.005 M I2 (0.01N), Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 1 Pc. |
| UN Number | NONH for all modes of transport |
| Beilstein | 3587194 |
| Packaging | Ampoule |
| Chemical Name or Material | Iodide Iodate concentrate |
Safety and Handling
| Hazard Statement | P305 + P351 + P338 |