missing translation for 'onlineSavingsMsg'
Learn More

Iodide Iodate concentrate, For 1L standard solution, 0.005 M I2 (0.01N), Honeywell Fluka™

Catalog No. 6002665
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002665 1 Pc.
60046687 6 x 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002665 Supplier Honeywell Chemicals Supplier No. 380651EA
Only null left

for 1L standard solution, 0.005 M I2 (0.01N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 153507-24-5
MDL Number MFCD00011355

Specifications

Quantity 1 Pc.
UN Number NONH for all modes of transport
Beilstein 3587194
Packaging Ampoule
Chemical Name or Material Iodide Iodate concentrate
Hazard Statement P305 + P351 + P338