Learn More
Hach™ IO9001 Module
Catalog No. 029380001
Change view
Click to view available options
Includes:
1 High Voltage Relay
Catalog No. Includes
029380001 1 High Voltage Relay
1 options
Catalog No. 029380001 Supplier Hach™ Supplier No. 02 93 800 01
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Adds versatility to the AS950 Automatic Sampler, connects through auxiliary port
  • Available for simple applications that only require 1 high voltage relay
  • For a broader range of inputs and outputs, see the IO9004 Module
Specifications
Product Type IO9001 Module
Description Connects through Auxillary Port
Includes 1 High Voltage Relay
For Use With (Application) AS950 Automatic Sampler