missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ DEPC-Treated Water
Autoclaved to remove the DEPC and 0.1-μm membrane-filtered.
$125.00 - $165.00
Specifications
| Grade | Molecular Biology |
|---|---|
| Treatment(s) | DEPC-Treated |
| Purification Method | Autoclaved, Membrane-Filtered |
| Product Line | UltraPure |
Description
UltraPure™ DEPC-treated Water is suitable for use with RNA. It is prepared by incubating with 0.1% diethylpyrocarbonate (DEPC), and is then autoclaved to remove the DEPC. Sterile filtered.
Performance and Quality Testing: No DNase or RNase activity detected.
Order Info
Shipping Condition: Room Temperature
Specifications
| Molecular Biology | |
| DEPC-Treated | |
| Autoclaved, Membrane-Filtered | |
| UltraPure |
For Research Use Only. Not for use in diagnostic procedures.