missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrueGuide™ sgRNA Positive Control, HPRT1 (human)

Catalog No. A35524
Encompass
Change view
Click to view available options
Quantity:
3 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A35524 3 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A35524 Supplier Invitrogen™ Supplier No. A35524

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for the optimization of transfection and verification of editing efficiency in transfected cells.

Provides complete collection of validated positive controls for the TrueGuide Synthetic gRNA product line to help maximize editing performance. This positive control against HPRT1 targets intronic region and has achieved editing efficiencies of up to 65% in variety of human cell lines. HPRT1 control easily determines the optimal conditions for delivering CRISPR/Cas9 tools to cell type, allowing to move forward in editing experiments and minimize downstream effort to isolate and validate your edited clones.

Validated positive control sgRNA targeting HPRT1

Specifications

Content And Storage Delivered as dried RNA. Store dried RNA at room temperature; store resuspended RNA at -5 to -30°C. Resuspend in nuclease-free water or TE.
Format Tube
Control Type Positive Control
RNAi Type gRNA
Species Human
Product Line TrueGuide
Product Type gRNA
Quantity 3 nmol
Shipping Condition Room Temperature
I would like to calculate the molecular weight of TrueGuide Synthetic gRNA? What is the length of TrueGuide Synthetic gRNA (TrueGuide sgRNA)?

The length of TrueGuide sgRNA is typically 100 nt. Also, the exact molecular weight can also be found on the COA.

For Research Use Only. Not for use in diagnostic procedures.