missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO DFS Item

Displays bright fluorescence when bound to RNA and low background in gels. Invitrogen™ SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO is ideal for use with either formaldehyde/agarose or polyacrylamide gels.

$728.00 - $1420.00

Specifications

Content And Storage • Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye SYBR Green II
Shipping Condition Room Temperature
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S7564 DFS Item
View Documents
Invitrogen™
S7564
500 μL
Each for $728.00
Only null left
 
S7568 DFS Item
View Documents
Invitrogen™
S7568
1 mL
Each for $1,196.00
Only null left
 
S7586 DFS Item
View Documents
Invitrogen™
S7586
20 x 50 μL
Each for $1,420.00
Only null left
 
Description

Description

Sensitive nucleic acid gel stain that fluoresces brightly when bound to RNA and low background in gels. SYBR Green II RNA is ideal for use with either formaldehyde/agarose or polyacrylamide gels using laser scanners or standard UV transilluminators. 500μL stains 50 minigels.

Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

• Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.
Fluorescence
Room Temperature
RNA Gel Stain
In-Gel Detection
SYBR Green II
RNA
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.