missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Resolving Buffer

Catalog No. p-6779027
Encompass
Change view
Click to view available options
Quantity:
2 x 500 mL
5 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HC2212 2 x 500 mL
HC2215 5 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. HC2212 Supplier Invitrogen™ Supplier No. HC2212
Only null left

These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank

Recommended Storage

Room Temperature

Specifications

Content And Storage 2 x 500 mL (1 L total) SureCast Resolving Buffer packs.

Store at room temperature.
Quantity 2 x 500 mL
Shipping Condition Room Temperature
Product Line SureCast
Product Type Resolving Buffer

For Research Use Only. Not for use in diagnostic procedures.