missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Resolving Buffer

Catalog No. HC2215
Encompass
Change view
Click to view available options
Quantity:
2 x 500 mL
5 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HC2215 5 x 500 mL
HC2212 2 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. HC2215 Supplier Invitrogen™ Supplier No. HC2215

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank

Recommended Storage

Room Temperature

Warranty

Up to 12 months

Specifications

Content And Storage 5 x 500 mL (2.5 L total) SureCast Resolving Buffer packs.

Store at room temperature.
Quantity 5 x 500 mL
Shipping Condition Room Temperature
Product Line SureCast
Product Type Resolving Buffer
Do you offer the SureCast casting reagents separately?

The SureCast Handcast System and reagents are available both as a package or as standalones.

Can I reuse SureCast Glass Plates?

Yes, SureCastGlass Plates can be reused.They are up to 22 times more durable than Bio-Rad glass plates.

Why should I store all SureCast reagents at room temperature

Having everything at room temperature means not having to thaw reagents, thus saving time in gel casting.

For Research Use Only. Not for use in diagnostic procedures.