missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rhinohide™ Polyacrylamide Gel Strengthener Concentrate

Catalog No. R33400
Encompass
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R33400 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R33400 Supplier Invitrogen™ Supplier No. R33400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Strengthens gels for easier handling and resistance to tearing

  • Improves upon classic polyacrylamide gel technology, without adverse side effects
  • Recommended for low-percentage gels, large-format gels and gels subject to multiple staining and handling steps
  • Available in a kit (Cat. No. R-33410), which includes an acrylamide/bis-acrylamide mixture (37.5:1 ratio)

Gel Casting, Protein Gel Electrophoresis, Proteins, Expression, Isolation & Analysis

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature.
Quantity 200 mL
Shipping Condition Room Temperature
Product Line Rhinohide
Product Type Polyacrylamide Gel Strengthener Concentrate
I am staining my protein gel with multiple stains over several days. How do I prevent excessive gel tearing?

For gels that you are pouring yourself, add Rhinohide Polyacrylamide Gel Strengthener (Cat. Nos. R33400, R33410) to the formulation. Decrease the shaker speed/motion; only very gentle rocking is required. Slowly decant solutions during solution changes and avoid touching the gel as much as possible when pouring out solutions.

For Research Use Only. Not for use in diagnostic procedures.