missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse miR RNAi Select

Catalog No. 100002319
Encompass
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
100002319 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 100002319 Supplier Invitrogen™ Supplier No. 100002319

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Easily design effective RNAi molecules for reliable results

  • Using BLOCK-iT™ RNAi Designer, design Custom BLOCK-iT siRNA sequences for efficient and specific gene expression knockdowns in any organism
  • For enhanced target specificity and stability in serum, Stealth RNAi™ RNAi chemistry is available

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at room temperature.
RNAi Type siRNA
Product Type Mouse miR RNAi Select
Quantity 1 Each
Target Mouse