missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ M13 Reverse Primer

Catalog No. N53002
Encompass
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N53002 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N53002 Supplier Invitrogen™ Supplier No. N53002
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oligonucleotides complementary to a DNA template are necessary to prime DNA synthesis for sequencing reactions.

Oligonucleotides complementary to a DNA template are necessary to prime DNA synthesis for sequencing reactions. We offer a wide selection of Invitrogen single-strand primers that may be used in either single- or double-stranded sequencing protocols. All sequencing primers are non-phosphorylated and are supplied lyophilized.

M13 Reverse Primer features include:

  • Desalted and purified by gel filtration
  • Assayed for function in automated sequencing reactions
  • Supplied in quantity of 2 μg

Order Info

Shipping Conditions: Room Temperature

Specifications

Product Type Primer
Content And Storage • M13 Reverse Primer (2 μg)

Store in Freezer at –20°C.
Guaranteed stable for 6 months when properly stored.
Primer Length 17-mer
Primer Sequence 5 ́d[CAGGAAACAGCTATGAC]3 ́
Purification Method Desalted, Gel Filtration Purified
Shipping Condition Room Temperature
Primer M13
Quantity 2 μg
For Use With (Application) Sequencing Reactions
Form Lyophilized

For Research Use Only. Not for use in diagnostic procedures.