missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luria Broth Base (Miller's LB Broth Base), powder

Catalog No. p-4918428
Encompass
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12795027 500 g
12795084 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12795027 Supplier Invitrogen™ Supplier No. 12795027
Only null left

Luria broth (LB) is a popular, rich medium used to cultivate bacteria and to perform coliphage plaque assays.

Luria broth (LB) is a popular, rich medium used to cultivate bacteria and to perform coliphage plaque assays. The original formulation published in 1951 by Guiseppe Bertani to study phage plaque formation and named Lysogeny broth. It is also known as Lennox broth and Luria-Bertani broth.

Widely used LB medium formulations differ in salt concentration:
Miller: 10 g/L NaCl
Lennox: 5 g/L NaCl
Luria: 0.5 g/L NaCl

Luria Broth Base (Miller's formulation) is recommended for the general maintenance and growth of E. coli as it allows for fast cell expansion and supports high cell density. In cases where salt-sensitive antibiotics are being used for selection (such as Zeocin, tetracycline or chloramphenicol), Lennox formulation medium would be recommended.

Luria Broth Base formulation
Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, and 10 g NaCl.
To prepare 1 L of Miller-LB media, add 25 g of powder to 1 L distilled water and mix by vortexing. Adjust the pH to 7.0 with 5 N NaOH and sterilize by autoclaving for 25 minutes at 121°C.

Find the media format that fits your needs
We offer other ready-to-use media formulations, such as LB Broth (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301) or S.O.C recovery medium (Cat. No. 15544034).

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage • 500 g LB Dehydrated Media Powder

Store at room temperature.

Format Bottle
Media Type LB Broth Base (Miller's LB Broth Base)
Preparation Method Autoclave
Product Type Dehydrated Culture Media
Target Organism Class Coliforms
Quantity 500 g
Shipping Condition Room Temperature
Species Escherichia coli

For Research Use Only. Not for use in diagnostic procedures.