missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LB Agar (Lennox L Agar), powder

Catalog No. 22700025
Encompass
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22700025 500 g
22700041 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22700025 Supplier Invitrogen™ Supplier No. 22700025

LB Agar (Lennox L agar) is a solid agar media powder recommended for the general maintenance and growth of E. coli strains.

LB Agar (Lennox L agar) is a solid agar media powder recommended for the general maintenance and growth of E. coli strains. As E. coli is unable to digest agar but can absorb nutrition from the LB medium, single colonies can be easily isolated and selected. With addition of a selective antibiotic, only transformed E. coli with resistance to that antibiotic can grow. The Lennox formulation has lower levels of salts than Miller and so is preferred when using salt-sensitive antibiotics such as Zeocin, tetracycline, or chloramphenicol.

Lennox broth is a popular, rich medium used to cultivate bacteria and to perform coliphage plaque assays. The original formulation was published in 1951 by Guiseppe Bertani in his study of phage plaque formation and named as Lysogeny broth. It is also known as Lennox broth and Luria-Bertani broth.

Solid culture media have been a major turning point for bacteriology, introduced by Robert Koch in 1881 by adding liquid cultures in gelatin (later replaced by agar derived from seaweed as per Walter Hesse's discovery). This allowed plated bacteria to form visible colonies on a solid and smooth surface. Koch’s assistant, Julius Petri, designed a shallow dish with a lid where medium could be poured, cooled to solidity, and protected from contamination.

Widely used LB medium formulations differ in salt concentration:
• Miller: 10 g/L NaCl
• Lennox: 5 g/L NaCl
• Luria: 0.5 g/L NaCl

LB Agar (Lennox L agar) formulation and preparation
Formulation per liter: 10 g SELECT Peptone 140, 5 g SELECT Yeast Extract, 5 g NaCl, and 12 g SELECT Agar.
To prepare 1 L of LB Agar medium (1.2% final agar concentration), weigh 37 g of powder and add distilled water to 1 L. Adjust the pH value to 7.0 with 5 N NaOH and sterilize by autoclaving for 25 minutes at 121°C.

Primary applications
• Select using antibiotic transformed E. coli colony with constructed DNA plasmid
• Streaking and isolating bacteria cells on LB agar plate
• Blue-white screening using X-Gal or Bluo-gal and IPTG LB agar plates

Find the media format that you need
• We offer other ready-to-use media formulations, such as LB (Lennox) (Cat. No. 10855001), Terrific Broth (Cat. No. A1374301) or S.O.C recovery medium (Cat. No. 15544034).
• Explore our media additives: X-Gal (Cat. No. 15520034), Bluo-gal (Cat. No. 15519028), IPTG (Cat. No. 15529019).

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage • 500 g LB Agar Dehydrated Media Powder

Store at room temperature.

Format Bottle
Media Type LB Agar (Lennox L agar)
Preparation Method Autoclave
Product Type Dehydrated Culture Media
Target Organism Class Coliforms
Quantity 500 g
Shipping Condition Room Temperature
Species Escherichia coli
I need to buy media to prepare agar plates, which I will be using to grow colonies after transformation (molecular cloning). Which product would you recommend?

We offer two options:

The first is our formulation of LB agar (catalog number 22700-025), which has LB Media and agar mixed together. This comes in a 500g and 2.5 kg size. All you have to do is measure out, add water, and autoclave.

The second option is even easier. Our imMedia comes premeasured in pouches. We have three types: imMedia Amp Agar, Kan Agar and Zeo Agar for ampicillin, kanamycin and Zeocin-containing plates. All you have to do is empty the pouch, add water, heat and pour plates. No autoclaving is needed! Each pouch contains sufficient reagents for 8-10 plates.

imMedia Amp Agar, 20 pouches, Q601-20

imMedia Kan Agar, 20 pouches, Q611-20

imMedia Zeo Agar, 20 pouches, Q621-20

For added convenience, you should also check out our LB medium imMedia products: (Each pouch makes 200 mL of media)

imMedia Amp Liquid, 20 pouches, Q600-20

imMedia Kan Liquid, 20 pouches, Q610-20

imMedia Zeo Liquid, 20 pouches, Q620-20

How much LB Agar, powder (Lennox L agar) (Cat. No. 22700025, 2700041) do I need to prepare 1 liter of LB Agar?

For preparing 1 liter of LB Agar, you need 32 g of the LB Agar, powder (Lennox L agar).

For Research Use Only. Not for use in diagnostic procedures.