Learn More
Invitrogen™ High DNA Mass Ladder
Description
Includes
High DNAmass Ladder is supplied at 520 ng/4μL in 10mM Tris-HCl (pH 7.5), 1mM EDTA Volume: 200μL; For 50 applications
High DNA Mass Ladder consists of an equimolar mixture of six blunt-ended DNA fragments of 10, 6, 4, 3, 2, and 1Kb. Electrophoresis of 4μL of the High DNA Mass Ladder results in bands containing 200, 120, 80, 60, 40, and 20ng (520ng total) of DNA, respectively. Compatible with agarose gels. Includes tartrazine, xylene cyanol.
- Less waste, sustainable packaging
Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | • 200 µL High DNA Mass Ladder • 1 mL10X BlueJuice Gel Loading Buffer Store at -20°C. |
| Concentration | 0.13 μg/μL |
| Gel Type | Agarose |
| Ready to Load | No |
| Sample Loading Volume | 1 mL |
| Size Range | 1000 to 10000 bp |
| Volume (Metric) | 200 μL |
| Gel Compatibility | Agarose gel |
| No. of Reactions | 50 Applications |
| Quantity | 26 μg |
| Show More |
Frequently Asked Questions (FAQs)
Sequences of Invitrogen DNA and RNA ladders are proprietary.
Invitrogen DNA ladders contain linear dsDNA fragments.
Invitrogen DNA ladders are composed of double-stranded DNA fragments only.
Here are a few reasons why you might see smearing of the bands:
- The DNA was degraded. Avoid nuclease contamination of DNA standards.
- Too much DNA was loaded on the gel. Decrease the amount of DNA in the gel.
- The DNA was contaminated by protein. Remove proteins by phenol extraction before electrophoresis.
- For small DNA, the bands may have diffused during staining. Add the ethidium bromide before electrophoresis.
- For radiolabeled DNA, labeling was performed by nick translation. Label the DNA by replacement synthesis with T4 DNA polymerase or label the 5' end with T4 polynucleotide kinase.
- Improper electrophoresis conditions were used. Do not allow voltage to exceed ~20 V/cm. Maintain a temperature <30°C during electrophoresis. Check that the electrophoresis buffer used has sufficient buffering capacity.
- The DNA contained too much salt. Remove excess salt by ethanol precipitation before electrophoresis.
This can happen if the marker was heated. Please ensure that the ladders are not heated before use.
For Research Use Only. Not for use in diagnostic procedures.