missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ β-Gal Assay Kit

Catalog No. k145501
Encompass
Change view
Click to view available options
Quantity:
80 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K145501 80 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K145501 Supplier Invitrogen™ Supplier No. K145501
Only null left

Includes

10X PBS (phosphate buffered saline), lysis buffer, cleavage buffer, β-mercaptoethanol, ONPG, stop buffer.

Provides reagents required to quickly measure levels of active β-galactosidase expressed in cells transfected with plasmids expressing lacZgene

  • lacZis bacterial gene often used as reporter construct in eukaryotic transfection experiments
  • Gene product of lacZ, β-galactosidase, is resistant to proteolysis in cellular lysates, and its activity is easily assayed
  • β-galactosidase catalyzes hydrolysis of β-galactosidase such as ortho-nitrophenyl-D-galactopyranoside (ONPG)
  • Hydrolysis of ONPG to ONP anion produces bright yellow color with peak absorbance at 420nm that can be quantified using spectrophotometer
  • 10.7 gene expression
  • Colorimetric detection

Beta-Galactosidase Assays, Enzyme and Protein Activity Assays, Proteins, Expression, Isolation and Analysis, Reporter Assays

Specifications

Content And Storage The β-Gal Assay Kit contains 10X PBS (phosphate buffered saline), lysis buffer, cleavage buffer, β-mercaptoethanol, ONPG, stop buffer. Store ONPG at -20°C. All other components are stored at either room temperature or +4°C. All reagents are guaranteed stable for 6 months when properly stored.
Assay β-Gal (lacZ) Assay
Detection Method Colorimetric
Product Type Beta-Gal Assay Kit
Substrate ONPG
Quantity 80 mL
Target Beta-Galactosidase

For Research Use Only. Not for use in diagnostic procedures.