missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ NGS™ 0.8% Agarose Gels
GREENER_CHOICE

Catalog No. a25798
Encompass
Change view
Click to view available options
Quantity:
10 gels
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25798 10 gels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25798 Supplier Invitrogen™ Supplier No. A25798
Only null left

Designed to provide fast, convenient, and easy electrophoresis

E-Gel NGS 0.8% Agarose Gels are self-contained, pre-cast agarose gels designed to provide fast, convenient, and easy electrophoresis. Each E-Gel agarose gel is ready for use with agarose, electrodes, and SYBR Safe DNA gel stain packaged inside an 11-well, easy-to-open, UV- and blue-light transparent cassette. SYBR Safe stain can be viewed with blue light, which minimizes DNA damage and allows safe, real-time monitoring of DNA migration using the E-Gel Power Snap Electrophoresis System.

Easy access to the gel
The gel cassette is designed to open with a gel knife, so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis.

Running and viewing apparatus
E-Gel NGS gels require the E-Gel Power Snap Electrophoresis System for running and viewing the gel in real time.

Specifications

Compatible Ladders E-Gel™ 1 Kb Plus DNA Ladder
Content And Storage Store at room temperature.
Gel Percentage 0.8%
Gel Type E-Gel
Label or Dye SYBR Safe
Separation Range 800 to 10,000 bp
Wells 11-well
Quantity 10 gels
Well Design Single Comb
Product Type Agarose Gel

For Research Use Only. Not for use in diagnostic procedures.