missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SampleRack (for DynaMag™-2 magnet)

This removable plastic rack supplied with the DynaMag-2 magnet.

Supplier:  Invitrogen™ 12322D

Encompass

Catalog No. 12322D

Only null left

Includes:

One acrylic copolymer SampleRack. Holds 16 microcentrifuge tubes.


Description

Description

This removable plastic rack supplied with the DynaMag-2 magnet

  • Holds 16 x 1.5 mL micro-centrifuge tubes
  • Tubes are kept in SampleRack, before, during and after isolation
  • When handling multiple samples in parallel, use additional SampleRacks to increase capacity
  • With two racks, keep up to 16 tubes in same SampleRack throughout Dynabeads isolation procedure, moving rack on and off the magnet as needed
  • SampleRack can be placed on bench, in refrigerator or incubator

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Contains one acrylic copolymer SampleRack. Holds 16 microcentrifuge tubes.

Store at 4-30°C.
DynaMag
Room Temperature
DynaMag™-2
Each
Sample Rack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.