missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Sheep-Anti Mouse IgG

Catalog No. 11031
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11031 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11031 Supplier Invitrogen™ Supplier No. LS11031
Only null left

Includes

5mL Dynabeads Sheep anti-Mouse IgG coated with sheep polyclonal against mouse IgG1 and IgG2 (not IgG3).

Dynabeads™ Sheep anti-Mouse IgG binds monoclonal mouse IgG of your choice for specific cell isolation or depletion.

Dynabeads™ Sheep anti-Mouse IgG binds monoclonal mouse IgG of your choice for specific cell isolation or depletion. All cells from all species except from mouse can be isolated with this product, depending only on the specific antibody of choice.

• Depletion: cells can be depleted from a sample
• Positive isolation: cells can be positively isolated from a sample for molecular applications
• Negative isolation: cells can be negatively isolated from a PBMC sample by adding an antibody cocktail of mouse IgGs towards the unwanted cells. Compatible with flow cytometry.

Note: For a more universal binding of your mouse IgG of any subclass, please use Dynabeads™ Pan Mouse Ig (Cat. No. 110-41D or 110-42D).

Starting samples:
Whole blood, PBMC, buffy coat., tissue digests.

Order Info

Shipping Condition: Room Temperature

Specifications

Antibody Fragment Whole Antibody
Cell Type All cells from all species, except mouse
Clonality Polyclonal
Content And Storage This Product Contains: 5 mL Dynabeads™ Sheep anti-Mouse IgG coated with sheep polyclonal against mouse IgG1 and IgG2 (not IgG3).

Store at 2°C to 8°C.
Product Type Antibody Coated Bead
Dye Type Dynabeads™
Host Species Sheep
Isolation Technology Depletion, negative isolation or positive isolation
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Reactivity All species, except mouse
Sample Type PBMC, Tissue Digests, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species All species, except mouse
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.