missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ CD25 DFS Item

Isolate CD25+ T-cells directly from whole blood or PBMC. Dynabeads™ CD25 are ideal for depleting CD25+ T-cells and for direct positive isolation for downstream molecular analysis.

Supplier:  Invitrogen™ 11157D

Catalog No. 11157D


Only null left

Includes: 5mL Dynabeads CD25 coated with anti-CD25 monoclonal antibody

Description

Description

Isolate CD25+ T-cells directly from whole blood or PBMC. Dynabeads™ CD25 are ideal for depleting CD25+ T-cells and for direct positive isolation for downstream molecular analysis.

• Depletion of CD25+ T-cells
• Isolation of pure and viable CD25+ T-cells for molecular applications
• No columns required

Note: For isolation of human CD4+CD25+Foxp3+ regulatory T-cells (Treg cells), please use Dynabeads™ Regulatory CD4+CD25+ T Cells (Cat. no. 113-63D).

Starting samples:
Whole blood, PBMC, buffy coat.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

T Cells (CD25)
Antibody Coated Bead
Processes ∼5 x 109 cells total
DYNAL, Dynabeads
5 mL
Room Temperature
Human
This Product Contains: 5 mL Dynabeads™ CD25 coated with anti-CD25 monoclonal antibody.

Store at 2°C to 8°C.
Depletion, positive isolation for molecular applications
>95%
Research Grade
PBMC, Blood
2.5 x 107 PBMCs per isolation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.