missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Exosome-Human CD81 Flow Detection Reagent (from cell culture)

Catalog No. 10622D
Encompass
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10622D 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10622D Supplier Invitrogen™ Supplier No. 10622D

Cell culture media must first be enriched for exosomes using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation.

Cell culture media must first be enriched for exosomes using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation. Exosome-Human CD81 Flow Detection Reagent (from cell culture) allows easy visualization of exosomes by flow cytometry using this magnetic separation technology. Free exosomes alone are too small to be detected by most flow cytometry instruments. The 2.7 μm Dynabeads magnetic beads allow for a clear and defined flow analysis.

  • Minimal hands-on time for specific exosome capture and analysis
  • Clear and defined flow analysis in less than 1 hour hands-on time
  • Study expression of specific exosome subset markers
  • Protocol is easily scalable
  • Recent studies suggest that exosome/ECV population consists of several subsets, possibly with different functions
  • Using immuno-affinity based Dynabeads magnetic separation technology, these subsets can be selectively captured and studied using flow cytometry and/or electron microscopy
  • Dynabeads technology avoids loss of material due to handling and centrifugation typically experienced with other isolation and analysis methods

Mix thoroughly to ensure that magnetic beads do not settle in the sample tube. A mixer which provides both tilting and rotation for optimal mixing is recommended.

Specifications

Content And Storage Store in refrigerator (2–8°C).
Product Type Flow Detection Reagent
Product Line Dynabeads
Quantity 2 mL
Sample Type Cell Cultures
Diameter (Metric) 2.7 μm

For Research Use Only. Not for use in diagnostic procedures.